Assay Details
Target Gene Details
Entrez Gene ID: | 6895 |
Gene Name: | TARBP2, RISC loading complex RNA binding subunit |
Gene Aliases: |
LOQS, TRBP, TRBP1, TRBP2 |
Location: |
Chr.12:53500921-53506431 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARBP2 | NM_004178.4 | NP_004169.3 | ||
| NM_134323.1 | NP_599150.1 | |||
| NM_134324.2 | NP_599151.2 | |||
| XM_005269114.1 | XP_005269171.1 | |||
| XM_005269115.2 | XP_005269172.1 | |||
| XM_005269117.1 | XP_005269174.1 | |||
| XM_005269120.4 | XP_005269177.1 | |||
| XM_005269122.3 | XP_005269179.1 | |||
| XM_006719581.1 | XP_006719644.1 | |||
| XM_011538712.2 | XP_011537014.1 | |||
| XM_017019910.1 | XP_016875399.1 | |||
| XM_017019911.1 | XP_016875400.1 | |||
| AK290737.1 | ||||
| BC005860.2 | AAH05860.1 | |||
| BI771121.1 | ||||
| BT007140.1 | AAP35804.1 | |||
| M60801.1 | AAA36765.1 | |||
| U08998.2 | AAB50581.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983486 | Chr.12:53503642 - 53514027 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map