Assay Details
Target Gene Details
Entrez Gene ID: | 84221 |
Gene Name: | spermatogenesis and centriole associated 1-like |
Gene Aliases: |
C21orf56 |
Location: |
Chr.21:46161148-46185159 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATC1L | NM_001142854.1 | 5 | 1856 | NP_001136326.1 |
| NM_032261.4 | 4 | 705 | NP_115637.3 | |
| XM_005261188.4 | 5 | 1631 | XP_005261245.1 | |
| XM_011529756.2 | 3 | 533 | XP_011528058.1 | |
| XM_017028480.1 | 6 | 1578 | XP_016883969.1 | |
| AK304535.1 | 5 | 1856 | ||
| AL136871.1 | 4 | 633 | CAB66805.1 | |
| BC009497.2 | 3 | 590 | ||
| BC065570.1 | 3 | 557 | ||
| BC084577.1 | 4 | 677 | AAH84577.1 | |
| BC093804.1 | 3 | 426 | ||
| BC112293.1 | 3 | 426 | ||
| CR533451.1 | 3 | 332 | CAG38482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587952 | Chr.21:46128641 - 46162188 on Build GRCh38 | Loss |
|
| nsv1061087 | Chr.21:46056174 - 46223305 on Build GRCh38 | Loss |
|
| nsv509804 | Chr.21:45879599 - 46235140 on Build GRCh38 | Insertion |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
| nsv470911 | Chr.21:46115334 - 46171856 on Build GRCh38 | Loss |
|
| nsv525728 | Chr.21:46052727 - 46163592 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map