Assay Details
Target Gene Details
Entrez Gene ID: | 1314 |
Gene Name: | coatomer protein complex subunit alpha |
Gene Aliases: |
AILJK, HEP-COP |
Location: |
Chr.1:160288587-160343564 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPA | NM_001098398.1 | NP_001091868.1 | ||
| NM_004371.3 | NP_004362.2 | |||
| AK307953.1 | ||||
| BC038447.1 | AAH38447.1 | |||
| U24105.1 | AAB70879.1 |
More Information
Set Membership: |
|

Genomic Map