Assay Details
Target Gene Details
Entrez Gene ID: | 11172 |
Gene Name: | insulin like 6 |
Gene Aliases: |
RIF1 |
Location: |
Chr.9:4992408-5185706 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INSL6 | XM_011517702.2 | XP_011516004.1 |
Target Gene Details
Entrez Gene ID: | 3717 |
Gene Name: | Janus kinase 2 |
Gene Aliases: |
JTK10, THCYT3 |
Location: |
Chr.9:4985086-5128183 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK2 | NM_001322194.1 | NP_001309123.1 | ||
| NM_001322195.1 | NP_001309124.1 | |||
| NM_001322196.1 | NP_001309125.1 | |||
| NM_001322198.1 | NP_001309127.1 | |||
| NM_001322199.1 | NP_001309128.1 | |||
| NM_004972.3 | NP_004963.1 | |||
| AF001362.1 | AAC23653.1 | |||
| AF005216.1 | AAB82092.1 | |||
| AF058925.1 | AAC23982.1 | |||
| AK292525.1 | ||||
| AY973034.1 | AAY22962.1 | |||
| BC039695.1 | AAH39695.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761481 | Chr.9:5024345 - 5040486 on Build GRCh38 | Loss |
|
| nsv527495 | Chr.9:5008070 - 5417881 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map