Assay Details
Target Gene Details
Entrez Gene ID: | 84955 |
Gene Name: | NudC domain containing 1 |
Gene Aliases: |
CML66, OVA66 |
Location: |
Chr.8:109240919-109334205 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDCD1 | NM_001128211.1 | 10 | 1594 | NP_001121683.1 |
| NM_032869.3 | 10 | 1694 | NP_116258.2 | |
| XM_017013910.1 | 11 | 1870 | XP_016869399.1 | |
| XM_017013911.1 | 11 | 1640 | XP_016869400.1 | |
| AF283301.1 | 10 | 1815 | AAK73017.1 | |
| AF283302.1 | 10 | 1552 | AAM69373.1 | |
| AF521133.1 | 10 | 1594 | AAQ08823.1 | |
| AK027897.1 | 10 | 1646 | BAB55439.1 | |
| AK301278.1 | 11 | 1710 | ||
| AK307663.1 | 10 | 1595 | ||
| AK315006.1 | 10 | 1661 | ||
| AL832317.1 | 7 | 1002 | CAD38612.1 | |
| BC000967.2 | 10 | 1639 | AAH00967.2 | |
| BC031258.1 | 11 | 1784 | AAH31258.1 | |
| BC043406.1 | 10 | 1659 | AAH43406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7281n100 | Chr.8:109157344 - 109290856 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map