Assay Details
Target Gene Details
Entrez Gene ID: | 79139 |
Gene Name: | derlin 1 |
Gene Aliases: |
DER-1, DER1 |
Location: |
Chr.8:123013164-123042423 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DERL1 | NM_001134671.2 | NP_001128143.1 | ||
| NM_024295.5 | NP_077271.1 | |||
| XM_006716657.1 | XP_006716720.1 | |||
| XM_011517299.1 | XP_011515601.1 | |||
| AK023846.1 | ||||
| AK124086.1 | ||||
| AK303826.1 | ||||
| AY358818.1 | AAQ89177.1 | |||
| BC002457.2 | AAH02457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032827 | Chr.8:122655842 - 123258160 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map