Assay Details
Target Gene Details
Entrez Gene ID: | 10994 |
Gene Name: | ilvB acetolactate synthase like |
Gene Aliases: |
209L8, AHAS, HACL1L, ILV2H |
Location: |
Chr.19:15114974-15125799 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILVBL | NM_006844.4 | 16 | 2132 | NP_006835.2 |
| XM_005259717.4 | 16 | 2279 | XP_005259774.1 | |
| XM_011527651.2 | 15 | 2111 | XP_011525953.1 | |
| AB209065.1 | 10 | 4059 | BAD92302.1 | |
| BC000109.1 | 5 | 754 | AAH00109.1 | |
| BC011722.2 | 16 | 2099 | AAH11722.1 | |
| BC011761.2 | 16 | 2025 | AAH11761.1 | |
| BM676342.1 | 1 | 216 | ||
| U61263.1 | 16 | 2007 | AAC50934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9668 | Chr.19:15101800 - 15131511 on Build GRCh38 | Gain |
|
| esv34154 | Chr.19:15101623 - 15284812 on Build GRCh38 | Loss |
|
| esv2758746 | Chr.19:14986138 - 15148368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map