Assay Details
Target Gene Details
Entrez Gene ID: | 84909 |
Gene Name: | chromosome 9 open reading frame 3 |
Gene Aliases: |
AOPEP, AP-O, APO, C90RF3, ONPEP |
Location: |
Chr.9:94726604-95148264 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621113 | Chr.9:94207958 - 94890837 on Build GRCh38 | Gain |
|
| nsv1041922 | Chr.9:94682694 - 94774471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map