Assay Details
Target Gene Details
Entrez Gene ID: | 8884 |
Gene Name: | solute carrier family 5 member 6 |
Gene Aliases: |
SMVT |
Location: |
Chr.2:27199587-27212307 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A6 | NM_021095.2 | 17 | 3031 | NP_066918.2 |
| NR_028323.1 | ||||
| XM_006712128.1 | 17 | 2826 | XP_006712191.1 | |
| XM_006712129.1 | 17 | 2874 | XP_006712192.1 | |
| XM_006712130.1 | 17 | 2865 | XP_006712193.1 | |
| XM_011533146.2 | 15 | 2591 | XP_011531448.1 | |
| XM_017005216.1 | 14 | 2601 | XP_016860705.1 | |
| AF069307.1 | 17 | 2927 | AAD31727.1 | |
| AF081571.1 | 16 | 2743 | AAD37481.1 | |
| AK027855.1 | ||||
| AK075078.1 | 8 | 1469 | BAC11387.1 | |
| AL096737.1 | 3 | 2085 | CAH56408.1 | |
| BC012806.2 | 15 | 2593 | AAH12806.1 | |
| BC015631.2 | 16 | 2744 | AAH15631.1 | |
| BG025980.1 | ||||
| N30242.1 | 1 | 242 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map