Assay Details
Target Gene Details
Entrez Gene ID: | 8825 |
Gene Name: | lin-7 homolog A, crumbs cell polarity complex component |
Gene Aliases: |
LIN-7A, LIN7, MALS-1, TIP-33, VELI1 |
Location: |
Chr.12:80792520-80937915 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIN7A | NM_001324423.1 | 5 | 893 | NP_001311352.1 |
| NM_004664.3 | 6 | 1012 | NP_004655.1 | |
| NR_136887.1 | 6 | 970 | ||
| NR_136888.1 | 7 | 1089 | ||
| XM_011538928.2 | 5 | 787 | XP_011537230.1 | |
| AF028826.1 | 5 | 728 | AAB84251.1 | |
| AF087693.1 | 6 | 981 | AAC78481.1 | |
| AF173081.1 | 6 | 1012 | AAD48500.1 | |
| BC099921.1 | 6 | 822 | ||
| BC118609.1 | 6 | 956 | ||
| BC122561.1 | 6 | 956 |
More Information
Set Membership: |
|

Genomic Map