Assay Details
Target Gene Details
Entrez Gene ID: | 5017 |
Gene Name: | ovo like transcriptional repressor 1 |
Gene Aliases: |
HOVO1 |
Location: |
Chr.11:65787034-65797219 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVOL1 | NM_004561.3 | 4 | 1958 | NP_004552.2 |
| XM_005274018.3 | 4 | 1776 | XP_005274075.1 | |
| XM_011545067.2 | 4 | 1774 | XP_011543369.1 | |
| XM_011545068.2 | 4 | 2319 | XP_011543370.1 | |
| XM_017017837.1 | 4 | 2612 | XP_016873326.1 | |
| XM_017017838.1 | 4 | 1651 | XP_016873327.1 | |
| BC059408.1 | 4 | 1934 | AAH59408.1 | |
| BX648725.1 | 4 | 1838 | CAH56127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map