Assay Details
Target Gene Details
Entrez Gene ID: | 411 |
Gene Name: | arylsulfatase B |
Gene Aliases: |
ASB, G4S, MPS6 |
Location: |
Chr.5:78777209-78986534 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSB | NM_000046.3 | 8 | 3021 | NP_000037.2 |
| XM_011543390.1 | 9 | 2234 | XP_011541692.1 | |
| J05225.1 | 8 | 1741 | AAA51784.1 | |
| M32373.1 | 8 | 2294 | AAA51779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762527 | Chr.5:78625376 - 79252373 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73770019] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map