Assay Details
Target Gene Details
Entrez Gene ID: | 1949 |
Gene Name: | ephrin B3 |
Gene Aliases: |
EFL6, EPLG8, LERK8 |
Location: |
Chr.17:7705202-7711375 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNB3 | NM_001406.3 | NP_001397.1 | ||
| AK027329.1 | ||||
| AK314841.1 | ||||
| BC022499.1 | AAH22499.1 | |||
| BC042944.2 | AAH42944.1 | |||
| U57001.1 | AAB05170.1 | |||
| U62775.1 | AAC50707.1 | |||
| U66406.1 | AAC51203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952119 | Chr.17:7700483 - 7709982 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map