Assay Details
Target Gene Details
Entrez Gene ID: | 79746 |
Gene Name: | enoyl-CoA hydratase domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.10:11742357-11764070 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECHDC3 | NM_024693.4 | 5 | 1231 | NP_078969.2 |
| XM_011519689.1 | 4 | 1005 | XP_011517991.1 | |
| AF275677.1 | 5 | 1205 | AAG24387.1 | |
| AF289604.1 | 6 | 1305 | AAL55788.1 | |
| AK024562.1 | 5 | 1144 | BAB14928.1 | |
| AK222511.1 | 5 | 1170 | BAD96231.1 | |
| AK290902.1 | 5 | 1161 | ||
| BC001091.1 | 5 | 964 | AAH01091.1 | |
| BC009617.1 | 5 | 1153 | AAH09617.1 |
More Information
Set Membership: |
|

Genomic Map