Assay Details
Target Gene Details
Entrez Gene ID: | 79741 |
Gene Name: | coiled-coil domain containing 7 |
Gene Aliases: |
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68 |
Location: |
Chr.10:32446082-32882864 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622830 | Chr.10:32468577 - 32479193 on Build GRCh38 | Loss |
|

Genomic Map