Assay Details
Target Gene Details
Entrez Gene ID: | 54788 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B12 |
Gene Aliases: |
DJ10 |
Location: |
Chr.10:72332830-72355149 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB12 | NM_001002762.2 | 8 | 3323 | NP_001002762.2 |
| NM_017626.4 | 9 | 2161 | NP_060096.3 | |
| XM_005269931.1 | 9 | 1958 | XP_005269988.1 | |
| XM_005269932.3 | 9 | 2006 | XP_005269989.1 | |
| AL833358.1 | 1 | 1392 |

Genomic Map