Assay Details
Target Gene Details
Entrez Gene ID: | 101054525 |
Gene Name: | progesterone receptor antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.11:101129077-101159270 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101054525 | NR_073144.1 | 1 | 635 | |
| FJ515872.1 | 1 | 635 | ||
| FJ515873.1 | 1 | 433 |
Target Gene Details
Entrez Gene ID: | 5241 |
Gene Name: | progesterone receptor |
Gene Aliases: |
NR3C3, PR |
Location: |
Chr.11:101029624-101130681 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGR | NM_000926.4 | 1 | 103 | NP_000917.3 |
| NM_001271162.1 | NP_001258091.1 | |||
| XM_006718858.3 | 1 | 953 | XP_006718921.1 | |
| XM_011542869.2 | 1 | 971 | XP_011541171.1 | |
| AK304853.1 | ||||
| X51730.1 | 1 | 814 | CAA36018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556131 | Chr.11:101115046 - 101609286 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map