Assay Details
Target Gene Details
Entrez Gene ID: | 401262 |
Gene Name: | cysteine rich protein 3 |
Gene Aliases: |
TLP, TLP-A, bA480N24.2 |
Location: |
Chr.6:43305473-43310705 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRIP3 | NM_206922.2 | NP_996805.2 | ||
| XM_005249103.3 | XP_005249160.1 | |||
| XM_011514609.2 | XP_011512911.1 | |||
| XM_017010856.1 | XP_016866345.1 | |||
| XM_017010857.1 | XP_016866346.1 | |||
| AY555741.1 | AAS66887.1 | |||
| AY555742.1 | AAS66888.1 | |||
| AY555743.1 | AAS66889.1 | |||
| AY555744.1 | AAS66890.1 | |||
| CN282994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602999 | Chr.6:43270871 - 43320155 on Build GRCh38 | Loss |
|
| nsv602998 | Chr.6:43245739 - 43308652 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv950170 | Chr.6:43275063 - 43310262 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map