Assay Details
Target Gene Details
Entrez Gene ID: | 285973 |
Gene Name: | autophagy related 9B |
Gene Aliases: |
APG9L2, NOS3AS, SONE |
Location: |
Chr.7:151012209-151024499 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG9B | NR_073169.1 | 17 | 2514 | |
| NR_133652.1 | 16 | 3251 | ||
| AY316116.1 | 11 | 1589 | AAQ86941.1 | |
| AY515311.1 | 16 | 3251 | AAS87212.1 | |
| BC128587.1 | 17 | 2480 |
Target Gene Details
Entrez Gene ID: | 4846 |
Gene Name: | nitric oxide synthase 3 |
Gene Aliases: |
ECNOS, eNOS |
Location: |
Chr.7:150991056-151014599 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOS3 | NM_000603.4 | 26 | 3684 | NP_000594.2 |
| XM_017012232.1 | 25 | 3441 | XP_016867721.1 | |
| XM_017012233.1 | 25 | 3340 | XP_016867722.1 | |
| AF400594.1 | 25 | 3388 | AAK83389.1 | |
| AK223636.1 | 25 | 3408 | BAD97356.1 | |
| AK225827.1 | 25 | 3408 | ||
| AK292928.1 | 25 | 3441 | ||
| AK300390.1 | 23 | 3115 | ||
| AK302686.1 | 25 | 3342 | ||
| AK315213.1 | 25 | 3410 | ||
| BC063294.1 | 26 | 3680 | AAH63294.1 | |
| BC069465.1 | 25 | 3406 | AAH69465.1 | |
| BU731828.1 | ||||
| M93718.1 | 25 | 3434 | AAA36364.1 | |
| M95296.1 | 25 | 3408 | AAA36372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map