Assay Details
Target Gene Details
Entrez Gene ID: | 81555 |
Gene Name: | Yip1 domain family member 5 |
Gene Aliases: |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
Location: |
Chr.5:144158159-144170714 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF5 | NM_001024947.3 | 5 | 980 | NP_001020118.1 |
| NM_001271732.1 | 4 | 860 | NP_001258661.1 | |
| NM_030799.8 | 5 | 729 | NP_110426.4 | |
| AB014733.1 | BAB20270.1 | |||
| AF140225.1 | 4 | 539 | AAG48521.1 | |
| AF318329.1 | 5 | 618 | AAL55836.1 | |
| AK054576.1 | 5 | 666 | BAB70763.1 | |
| AK124065.1 | 5 | 887 | ||
| AK223515.1 | 5 | 592 | BAD97235.1 | |
| AK307251.1 | 5 | 348 | ||
| AY037152.1 | 5 | 661 | AAK67644.1 | |
| AY358863.1 | 5 | 613 | AAQ89222.1 | |
| AY640925.1 | 5 | 645 | AAV51256.1 | |
| AY640926.1 | 5 | 645 | AAV51257.1 | |
| AY640927.1 | 5 | 645 | AAV51258.1 | |
| AY640928.1 | 5 | 896 | AAV51259.1 | |
| AY640929.1 | 4 | 776 | ||
| AY640934.1 | 5 | 645 | AAV51260.1 | |
| BC007829.2 | 5 | 647 | AAH07829.1 | |
| BC014253.2 | 5 | 667 | AAH14253.1 | |
| BC024737.1 | 5 | 664 | AAH24737.1 | |
| BX647859.1 | 5 | 669 | ||
| CR749463.1 | 5 | 668 | CAH18295.1 |
More Information
Set Membership: |
|

Genomic Map