Assay Details
Target Gene Details
Entrez Gene ID: | 54752 |
Gene Name: | fibronectin type III domain containing 8 |
Gene Aliases: |
- |
Location: |
Chr.17:35121612-35130732 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC8 | NM_017559.2 | 1 | 48 | NP_060029.1 |
| AK225743.1 | ||||
| AK313775.1 | 1 | 81 | ||
| AL133054.1 | CAB61380.1 | |||
| BC024002.2 | AAH24002.1 | |||
| BI829497.1 | 1 | 48 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|

Genomic Map