Assay Details
Target Gene Details
Entrez Gene ID: | 727993 |
Gene Name: | uncharacterized LOC727993 |
Gene Aliases: |
- |
Location: |
Chr.20:1947210-2007517 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC727993 | NR_134520.1 | |||
| AK090681.1 |
Target Gene Details
Entrez Gene ID: | 5173 |
Gene Name: | prodynorphin |
Gene Aliases: |
ADCA, PENKB, SCA23 |
Location: |
Chr.20:1978756-1994285 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDYN | NM_001190892.1 | 3 | 1714 | NP_001177821.1 |
| NM_001190898.2 | 4 | 1923 | NP_001177827.1 | |
| NM_001190899.2 | 3 | 1866 | NP_001177828.1 | |
| NM_001190900.1 | 4 | 1613 | NP_001177829.1 | |
| NM_024411.4 | 4 | 1926 | NP_077722.1 | |
| XM_011529244.1 | 3 | 1606 | XP_011527546.1 | |
| XM_011529245.1 | 3 | 1797 | XP_011527547.1 | |
| XM_011529246.2 | 3 | 1784 | XP_011527548.1 | |
| XM_011529247.1 | 3 | 1593 | XP_011527549.1 | |
| XM_011529248.1 | 3 | 1706 | XP_011527550.1 | |
| XM_011529249.2 | 3 | 2336 | XP_011527551.1 | |
| XM_011529250.2 | 3 | 1747 | XP_011527552.1 | |
| XM_017027878.1 | 3 | 1596 | XP_016883367.1 | |
| AK295512.1 | 2 | 1188 | ||
| BC026334.1 | 4 | 1699 | AAH26334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map