Assay Details
Target Gene Details
Entrez Gene ID: | 1479 |
Gene Name: | cleavage stimulation factor subunit 3 |
Gene Aliases: |
CSTF-77 |
Location: |
Chr.11:33084584-33161491 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSTF3 | NM_001326.2 | 21 | 2557 | NP_001317.1 |
| AK290836.1 | 21 | 2557 | ||
| AK294204.1 | 18 | 2150 | ||
| AK297371.1 | 20 | 2439 | ||
| BC010533.1 | 6 | 989 | AAH10533.1 | |
| BC108319.1 | 21 | 2531 | AAI08320.1 | |
| U15782.1 | 21 | 2522 | AAA61417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1093n100 | Chr.11:33007427 - 33477800 on Build GRCh38 | Gain |
|
| esv2762904 | Chr.11:32947712 - 33564989 on Build GRCh38 | Gain |
|
| nsv521130 | Chr.11:33046843 - 33581367 on Build GRCh38 | Gain |
|
| dgv1742n54 | Chr.11:33043848 - 33455157 on Build GRCh38 | Gain |
|
| nsv975378 | Chr.11:33083645 - 33091064 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map