Assay Details
Target Gene Details
Entrez Gene ID: | 58487 |
Gene Name: | CREB/ATF bZIP transcription factor |
Gene Aliases: |
SMILE, ZF |
Location: |
Chr.11:85657564-85665148 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBZF | NM_001039618.2 | 1 | 3070 | NP_001034707.1 |
| NR_028024.1 | 2 | 2004 | ||
| NR_028025.1 | 3 | 1791 | ||
| NR_028026.1 | ||||
| NR_028027.1 | ||||
| XM_006718642.3 | 3 | 2637 | XP_006718705.1 | |
| XM_011545195.2 | 2 | 2850 | XP_011543497.1 | |
| XM_017018086.1 | 4 | 1916 | XP_016873575.1 | |
| XM_017018087.1 | XP_016873576.1 | |||
| XM_017018088.1 | 2 | 2359 | XP_016873577.1 | |
| XM_017018089.1 | XP_016873578.1 | |||
| XM_017018090.1 | 3 | 2129 | XP_016873579.1 | |
| XM_017018091.1 | XP_016873580.1 | |||
| XM_017018092.1 | XP_016873581.1 | |||
| AF039942.1 | 2 | 1952 | AAD28325.1 | |
| BC060807.1 | 1 | 3061 | ||
| BQ720558.1 | ||||
| CA843387.1 | ||||
| CN275782.1 | ||||
| DQ128105.1 | 2 | 1522 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759845 | Chr.11:85469436 - 85892945 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78958047] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map