Assay Details
Target Gene Details
Entrez Gene ID: | 63916 |
Gene Name: | engulfment and cell motility 2 |
Gene Aliases: |
CED-12, CED12, Ced-12A, ELMO-2 |
Location: |
Chr.20:46366045-46407051 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO2 | NM_001318253.1 | 22 | 3759 | NP_001305182.1 |
| NM_133171.4 | 22 | 3691 | NP_573403.1 | |
| NM_182764.2 | 21 | 3587 | NP_877496.1 | |
| XM_005260496.3 | 22 | 3256 | XP_005260553.1 | |
| XM_005260498.3 | 21 | 3618 | XP_005260555.1 | |
| XM_005260499.3 | 21 | 3589 | XP_005260556.1 | |
| XM_005260500.3 | 20 | 3117 | XP_005260557.1 | |
| XM_005260501.4 | 21 | 3127 | XP_005260558.1 | |
| XM_006723854.3 | 21 | 3190 | XP_006723917.1 | |
| XM_017028009.1 | 21 | 3562 | XP_016883498.1 | |
| XM_017028010.1 | 20 | 3065 | XP_016883499.1 | |
| XM_017028011.1 | 20 | 3518 | XP_016883500.1 | |
| AB058737.1 | 21 | 3202 | ||
| AF417861.1 | 20 | 3062 | AAL38512.1 | |
| AK021718.1 | BAB13879.1 | |||
| AK022731.1 | 9 | 1973 | BAB14210.1 | |
| AK023103.1 | 10 | 2018 | BAB14405.1 | |
| AK092796.1 | 2 | 1767 | ||
| AL713765.1 | 10 | 3704 | ||
| AL833406.1 | 21 | 3168 | ||
| BC000143.4 | 21 | 3267 | AAH00143.2 | |
| BX648819.1 | 21 | 3168 | ||
| CA444293.1 | 1 | 435 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586155 | Chr.20:46340501 - 46387008 on Build GRCh38 | Loss |
|
| nsv1057686 | Chr.20:46105657 - 46434770 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map