Assay Details
Target Gene Details
Entrez Gene ID: | 10980 |
Gene Name: | COP9 signalosome subunit 6 |
Gene Aliases: |
CSN6, MOV34-34KD |
Location: |
Chr.7:100088960-100092200 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS6 | NM_006833.4 | NP_006824.2 | ||
| AK295237.1 | 3 | 293 | ||
| AK312024.1 | ||||
| BC002520.2 | AAH02520.2 | |||
| BI113801.1 | ||||
| CB990467.1 | ||||
| U70735.1 | AAD03469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map