Assay Details
Target Gene Details
Entrez Gene ID: | 348 |
Gene Name: | apolipoprotein E |
Gene Aliases: |
AD2, APO-E, LDLCQ5, LPG |
Location: |
Chr.19:44905749-44909395 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOE | NM_000041.3 | 4 | 1007 | NP_000032.1 |
| NM_001302688.1 | 4 | 1089 | NP_001289617.1 | |
| NM_001302689.1 | 4 | 938 | NP_001289618.1 | |
| NM_001302690.1 | 4 | 1038 | NP_001289619.1 | |
| NM_001302691.1 | 4 | 1022 | NP_001289620.1 | |
| AK314898.1 | 4 | 960 | ||
| AL037046.3 | 1 | 162 | ||
| BC003557.1 | 4 | 936 | AAH03557.1 | |
| K00396.1 | 4 | 951 | AAB59546.1 | |
| KU177910.1 | 3 | 891 | ||
| KU177911.1 | 4 | 507 | ||
| KU177912.1 | 3 | 324 | ||
| KU177913.1 | 3 | 315 | ||
| M12529.1 | 4 | 952 | AAB59518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509747 | Chr.19:44877936 - 44943426 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map