Assay Details
Target Gene Details
Entrez Gene ID: | 2823 |
Gene Name: | glycoprotein M6A |
Gene Aliases: |
GPM6, M6A |
Location: |
Chr.4:175632937-176002691 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPM6A | NM_001261447.1 | 5 | 2114 | NP_001248376.1 |
| NM_001261448.1 | 7 | 2422 | NP_001248377.1 | |
| NM_005277.4 | 8 | 2523 | NP_005268.1 | |
| NM_201591.2 | 7 | 3005 | NP_963885.1 | |
| NM_201592.2 | 7 | 2464 | NP_963886.1 | |
| NR_048571.1 | 6 | 2173 | ||
| XM_006714183.1 | 6 | 2093 | XP_006714246.1 | |
| XM_011531877.1 | 7 | 2362 | XP_011530179.1 | |
| XM_017008036.1 | 7 | 2416 | XP_016863525.1 | |
| AK226176.1 | 7 | 2156 | ||
| BC022508.1 | 7 | 2155 | AAH22508.1 | |
| BC022528.1 | 8 | 2165 | AAH22528.1 | |
| BC037853.1 | 3 | 2498 | AAH37853.1 | |
| BC044612.1 | 7 | 2635 | AAH44612.1 | |
| D49958.1 | 7 | 2211 | BAA08712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470094 | Chr.4:175537097 - 175649860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map