Assay Details
Target Gene Details
Entrez Gene ID: | 51406 |
Gene Name: | nucleolar protein 7 |
Gene Aliases: |
C6orf90, PQBP3, RARG-1, dJ223E5.2 |
Location: |
Chr.6:13615327-13632470 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL7 | NM_001317724.1 | NP_001304653.1 | ||
| BU071935.1 |
Target Gene Details
Entrez Gene ID: | 10048 |
Gene Name: | RAN binding protein 9 |
Gene Aliases: |
BPM-L, BPM90, RANBPM, RanBP7 |
Location: |
Chr.6:13621498-13711582 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP9 | NM_005493.2 | NP_005484.2 | ||
| XM_006714945.2 | XP_006715008.1 | |||
| XM_011514205.2 | XP_011512507.1 | |||
| XM_017010149.1 | XP_016865638.1 | |||
| AB008515.1 | BAA23216.1 | |||
| AB055311.1 | BAB62525.1 | |||
| AF306510.1 | AAK15469.1 | |||
| AK313334.1 | ||||
| BC052781.1 | AAH52781.1 | |||
| BC063849.1 | AAH63849.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019422 | Chr.6:13621911 - 13694549 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map