Assay Details
Target Gene Details
Entrez Gene ID: | 57158 |
Gene Name: | junctophilin 2 |
Gene Aliases: |
CMH17, JP-2, JP2 |
Location: |
Chr.20:44111697-44187578 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JPH2 | NM_020433.4 | 6 | 4275 | NP_065166.2 |
| AK098288.1 | 1 | 1797 | ||
| AK098303.1 | 1 | 1797 | ||
| AW014086.1 | ||||
| BC043206.2 | 5 | 2565 | AAH43206.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv954043 | Chr.20:44109461 - 44117660 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map