Assay Details
Target Gene Details
Entrez Gene ID: | 79947 |
Gene Name: | dehydrodolichyl diphosphate synthase subunit |
Gene Aliases: |
CIT, CPT, DS, HDS, RP59 |
Location: |
Chr.1:26432282-26471306 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHDDS | NM_001243564.1 | 1 | 42 | NP_001230493.1 |
| NM_001243565.1 | 1 | 42 | NP_001230494.1 | |
| NM_001319959.1 | 1 | 42 | NP_001306888.1 | |
| NM_024887.3 | 1 | 42 | NP_079163.2 | |
| NM_205861.2 | 1 | 42 | NP_995583.1 | |
| XM_006710915.2 | XP_006710978.1 | |||
| XM_006710917.2 | XP_006710980.1 | |||
| XM_017002384.1 | XP_016857873.1 | |||
| AK023164.1 | BAB14439.1 | |||
| AK308187.1 | ||||
| AK316485.1 | ||||
| BC004117.2 | AAH04117.1 | |||
| BX648507.1 | 1 | 44 | ||
| DA756178.1 |
More Information
Set Membership: |
|

Genomic Map