Assay Details
Target Gene Details
Entrez Gene ID: | 100129620 |
Gene Name: | uncharacterized LOC100129620 |
Gene Aliases: |
- |
Location: |
Chr.1:99004276-99148852 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129620 | NR_033940.1 | 1 | 320 | |
| AK095544.1 | 1 | 320 | BAC04569.1 |
Target Gene Details
Entrez Gene ID: | 163404 |
Gene Name: | phospholipid phosphatase related 5 |
Gene Aliases: |
LPPR5, PAP2, PAP2D, PRG5 |
Location: |
Chr.1:98890245-99005850 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPPR5 | NM_001010861.2 | 1 | 299 | NP_001010861.1 |
| NM_001037317.1 | 1 | 299 | NP_001032394.1 | |
| XM_011540836.2 | 1 | 293 | XP_011539138.1 | |
| XM_011540837.1 | 1 | 293 | XP_011539139.1 | |
| XM_011540838.2 | XP_011539140.1 | |||
| AK295688.1 | ||||
| AY574039.1 | 1 | 254 | AAS76645.1 | |
| AY634620.1 | 1 | 132 | AAT64918.1 | |
| BC040174.1 | AAH40174.1 | |||
| BC136350.1 | 1 | 237 | ||
| BC144000.1 | 1 | 237 | ||
| BF514931.1 | 1 | 396 | ||
| DR001828.1 | 1 | 303 | ||
| FJ472844.1 | 1 | 299 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546884 | Chr.1:98987731 - 99062363 on Build GRCh38 | Loss |
|
| nsv830771 | Chr.1:98877965 - 99049970 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map