Assay Details
Target Gene Details
Entrez Gene ID: | 11185 |
Gene Name: | indolethylamine N-methyltransferase |
Gene Aliases: |
TEMT |
Location: |
Chr.7:30752135-30757602 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INMT | NM_001199219.1 | 3 | 426 | NP_001186148.1 |
| NM_006774.4 | 3 | 429 | NP_006765.4 | |
| AF128846.1 | 3 | 429 | AAF18304.1 | |
| AF128847.1 | 3 | 429 | AAF18305.1 | |
| AK309519.1 | 4 | 559 | ||
| AK313832.1 | 3 | 424 | ||
| BC033813.1 | 3 | 411 | AAH33813.1 | |
| BC103712.1 | 3 | 412 | AAI03713.1 | |
| BC106902.1 | 3 | 429 | AAI06903.1 | |
| BC106903.1 | 3 | 429 | AAI06904.1 | |
| DA920005.1 | 3 | 429 |
Target Gene Details
Entrez Gene ID: | 100526825 |
Gene Name: | INMT-FAM188B readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.7:30752135-30892387 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INMT-FAM188B | NR_037598.1 | |||
| AK308762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606506 | Chr.7:30730693 - 30760082 on Build GRCh38 | Loss |
|
| dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain |
|
| nsv830938 | Chr.7:30644614 - 30857907 on Build GRCh38 | Gain |
|
| nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain |
|
| esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain |
|
| nsv830939 | Chr.7:30704855 - 30871891 on Build GRCh38 | Loss |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map