Assay Details
Target Gene Details
Entrez Gene ID: | 27292 |
Gene Name: | DIM1 dimethyladenosine transferase 1 homolog |
Gene Aliases: |
DIM1, DIMT1L, HSA9761, HUSSY5 |
Location: |
Chr.5:62388523-62403939 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIMT1 | NM_014473.2 | NP_055288.1 | ||
| XM_011543323.2 | XP_011541625.1 | |||
| AF091078.1 | AAC72947.1 | |||
| AF102147.1 | AAC97955.1 | |||
| AJ009761.1 | CAA08815.1 | |||
| AK292723.1 | ||||
| BC002841.2 | AAH02841.1 | |||
| BC010874.2 | AAH10874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5689n100 | Chr.5:62155965 - 62735538 on Build GRCh38 | Gain |
|
| esv35035 | Chr.5:62362116 - 62496416 on Build GRCh38 | Gain |
|
| esv3605233 | Chr.5:62352619 - 62395105 on Build GRCh38 | Gain |
|
| dgv5690n100 | Chr.5:62310783 - 62615326 on Build GRCh38 | Gain |
|
| esv3894198 | Chr.5:62338170 - 62527080 on Build GRCh38 | Gain |
|
| nsv598348 | Chr.5:62355016 - 62599297 on Build GRCh38 | Gain |
|
| esv2759345 | Chr.5:62107311 - 62453085 on Build GRCh38 | Gain+Loss |
|
| esv3605231 | Chr.5:62328191 - 62537641 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map