Assay Details
Target Gene Details
Entrez Gene ID: | 286336 |
Gene Name: | family with sequence similarity 78 member A |
Gene Aliases: |
C9orf59 |
Location: |
Chr.9:131258078-131281930 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM78A | NM_033387.3 | 2 | 3191 | NP_203745.2 |
| XM_011518567.1 | 3 | 2734 | XP_011516869.1 | |
| XM_011518568.2 | 2 | 3437 | XP_011516870.1 | |
| AK024434.1 | 3 | 3813 | BAB15724.1 | |
| BC029924.2 | 2 | 2605 | AAH29924.2 | |
| BC049388.2 | 2 | 3191 | AAH49388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509327 | Chr.9:131241381 - 131448812 on Build GRCh38 | Insertion |
|
| nsv951790 | Chr.9:131256914 - 131262513 on Build GRCh38 | Deletion |
|
| nsv831739 | Chr.9:131145346 - 131310794 on Build GRCh38 | Loss |
|

Genomic Map