Assay Details
Target Gene Details
Entrez Gene ID: | 342574 |
Gene Name: | keratin 27 |
Gene Aliases: |
K25IRS3, KRT25C |
Location: |
Chr.17:40776808-40782534 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT27 | NM_181537.3 | NP_853515.2 | ||
| AJ564206.1 | CAD91906.1 |
More Information
Set Membership: |
|

Genomic Map