Assay Details
Target Gene Details
Entrez Gene ID: | 85315 |
Gene Name: | progestin and adipoQ receptor family member 8 |
Gene Aliases: |
C6orf33, LMPB1, MPRB |
Location: |
Chr.6:52362128-52407777 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR8 | NM_133367.4 | 2 | 2303 | NP_588608.1 |
| AF313619.1 | 1 | 1735 | AAO47232.1 | |
| AF347029.1 | 2 | 2293 | AAL79777.2 | |
| AK057083.1 | 1 | 338 | ||
| AK093603.1 | 1 | 1919 | ||
| AK128085.1 | 1 | 2523 | ||
| AY424286.1 | 3 | 2306 | AAR08374.1 | |
| BC030664.2 | 2 | 2278 | AAH30664.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608996 | Chr.6:52379078 - 52424700 on Build GRCh38 | Gain |
|
| nsv1025395 | Chr.6:52380188 - 52438563 on Build GRCh38 | Gain |
|
| nsv1027645 | Chr.6:52376160 - 52417602 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map