Assay Details
Target Gene Details
Entrez Gene ID: | 3595 |
Gene Name: | interleukin 12 receptor subunit beta 2 |
Gene Aliases: |
- |
Location: |
Chr.1:67307351-67397730 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12RB2 | NM_001258214.1 | NP_001245143.1 | ||
| NM_001258215.1 | NP_001245144.1 | |||
| NM_001258216.1 | NP_001245145.1 | |||
| NM_001319233.1 | NP_001306162.1 | |||
| NM_001559.2 | NP_001550.1 | |||
| NR_047583.1 | ||||
| NR_047584.1 | ||||
| XM_005270825.2 | XP_005270882.1 | |||
| XM_005270827.2 | XP_005270884.1 | |||
| XM_005270828.3 | XP_005270885.1 | |||
| XM_006710617.2 | XP_006710680.1 | |||
| XM_011541383.2 | XP_011539685.1 | |||
| XM_011541384.2 | XP_011539686.1 | |||
| XM_017001202.1 | XP_016856691.1 | |||
| XM_017001203.1 | XP_016856692.1 | |||
| XM_017001204.1 | XP_016856693.1 | |||
| AK294494.1 | ||||
| AK316525.1 | ||||
| BC104772.1 | AAI04773.1 | |||
| BC104774.1 | AAI04775.1 | |||
| BC143249.1 | ||||
| U64198.1 | AAB36675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10228 | Chr.1:67326710 - 67330488 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map