Assay Details
Target Gene Details
Entrez Gene ID: | 252995 |
Gene Name: | fibronectin type III domain containing 5 |
Gene Aliases: |
FRCP2, irisin |
Location: |
Chr.1:32862268-32872492 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC5 | NM_001171940.1 | NP_001165411.2 | ||
| NM_001171941.2 | 5 | 590 | NP_001165412.1 | |
| NM_153756.2 | NP_715637.2 | |||
| AK092102.1 | BAC03806.1 | |||
| BC062297.1 | 5 | 579 | AAH62297.1 | |
| BX537781.1 | CAD97840.1 | |||
| DA393478.1 | 3 | 260 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516302 | Chr.1:32855000 - 32914507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map