Assay Details
Target Gene Details
Entrez Gene ID: | 445577 |
Gene Name: | chromosome 9 open reading frame 129 |
Gene Aliases: |
bA165J3.3 |
Location: |
Chr.9:93318199-93346414 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf129 | NM_001098808.1 | NP_001092278.1 | ||
| XM_006717117.3 | XP_006717180.1 |
Target Gene Details
Entrez Gene ID: | 65268 |
Gene Name: | WNK lysine deficient protein kinase 2 |
Gene Aliases: |
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43 |
Location: |
Chr.9:93184156-93327581 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNK2 | NM_006648.3 | NP_006639.3 | ||
| XM_005252141.2 | XP_005252198.1 | |||
| AK000694.1 | ||||
| AK093910.1 | ||||
| AK124314.1 | ||||
| BC037965.1 | AAH37965.1 |
More Information
Set Membership: |
|

Genomic Map