Assay Details
Target Gene Details
Entrez Gene ID: | 3744 |
Gene Name: | potassium voltage-gated channel subfamily A member 10 |
Gene Aliases: |
Kcn1, Kv1.8 |
Location: |
Chr.1:110517217-110519175 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA10 | NM_005549.2 | 1 | 1254 | NP_005540.1 |
| BC074990.2 | 1 | 896 | AAH74990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006346 | Chr.1:110477152 - 110600523 on Build GRCh38 | Gain |
|
| nsv547498 | Chr.1:110492552 - 110518440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map