Assay Details
Target Gene Details
Entrez Gene ID: | 10435 |
Gene Name: | CDC42 effector protein 2 |
Gene Aliases: |
BORG1, CEP2 |
Location: |
Chr.11:65314729-65322429 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP2 | NM_006779.3 | 2 | 1503 | NP_006770.1 |
| XM_017017094.1 | 2 | 1614 | XP_016872583.1 | |
| AF001436.1 | 2 | 1461 | AAB81206.1 | |
| AF163840.1 | 2 | 1461 | AAD48784.1 | |
| AK098292.1 | 2 | 1617 | ||
| BC022337.1 | 1 | 1098 | AAH22337.1 | |
| BC075834.1 | 2 | 1404 | AAH75834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422285 | Chr.11:65245823 - 65487069 on Build GRCh38 | Deletion |
|
| nsv832190 | Chr.11:65210673 - 65360142 on Build GRCh38 | Loss |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map