Assay Details
Target Gene Details
Entrez Gene ID: | 80237 |
Gene Name: | elongation factor for RNA polymerase II 3 |
Gene Aliases: |
- |
Location: |
Chr.15:43772600-43777304 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELL3 | NM_025165.2 | NP_079441.1 | ||
| AF276512.1 | AAG13463.1 | |||
| AK026290.1 | BAB15432.1 | |||
| AK057528.1 | ||||
| AK126384.1 | ||||
| BC006548.2 | AAH06548.1 | |||
| BC019293.1 | AAH19293.1 | |||
| CR457358.1 | CAG33639.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map