Assay Details
Target Gene Details
Entrez Gene ID: | 23395 |
Gene Name: | leucyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
LEURS, PRLTS4, mtLeuRS |
Location: |
Chr.3:45388561-45548842 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARS2 | NM_015340.3 | NP_056155.1 | ||
| XM_005265006.1 | XP_005265063.1 | |||
| XM_011533554.2 | XP_011531856.1 | |||
| XM_017006042.1 | XP_016861531.1 | |||
| AK025465.1 | ||||
| AK308146.1 | ||||
| BC025989.2 | AAH25989.1 | |||
| D21851.1 |
Target Gene Details
Entrez Gene ID: | 100885795 |
Gene Name: | LARS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:45482695-45509545 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARS2-AS1 | NR_048543.1 | |||
| DB017595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834677 | Chr.3:45343270 - 45511215 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map