Assay Details
Target Gene Details
Entrez Gene ID: | 64220 |
Gene Name: | stimulated by retinoic acid 6 |
Gene Aliases: |
MCOPCB8, MCOPS9, PP14296 |
Location: |
Chr.15:74179466-74212267 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map