Assay Details
Target Gene Details
Entrez Gene ID: | 127687 |
Gene Name: | chromosome 1 open reading frame 122 |
Gene Aliases: |
ALAESM |
Location: |
Chr.1:37807801-37809454 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf122 | NM_001142726.1 | 3 | 850 | NP_001136198.1 |
| NM_198446.2 | 3 | 983 | NP_940848.2 | |
| AK127381.1 | 3 | 487 | BAC86950.1 | |
| AY517499.1 | 3 | 638 | ||
| BC057819.1 | 3 | 488 | AAH57819.1 | |
| BC102041.1 | 2 | 139 | AAI02042.1 | |
| BC127187.1 | 2 | 139 | ||
| BC127188.1 | ||||
| CV575592.1 |

Genomic Map