Assay Details
Target Gene Details
Entrez Gene ID: | 92342 |
Gene Name: | methyltransferase like 18 |
Gene Aliases: |
AsTP2, C1orf156, HPM1 |
Location: |
Chr.1:169792524-169794976 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL18 | NM_001320199.1 | 2 | 575 | NP_001307128.1 |
| NM_001320201.1 | 2 | 525 | NP_001307130.1 | |
| NM_033418.3 | 2 | 630 | NP_219486.1 | |
| XM_006711627.3 | 2 | 1083 | XP_006711690.1 | |
| AK313909.1 | 2 | 549 | ||
| AL035369.1 | 2 | 544 | CAA23019.1 | |
| AL556604.3 | 2 | 620 | ||
| AY744366.1 | 1 | 266 | AAU95377.1 | |
| BC008679.1 | 2 | 525 | AAH08679.1 | |
| BG614785.1 | 2 | 501 | ||
| CR450330.1 | 1 | 266 | CAG29326.1 |
More Information
Set Membership: |
|

Genomic Map