Assay Details
Target Gene Details
Entrez Gene ID: | 5068 |
Gene Name: | regenerating family member 3 alpha |
Gene Aliases: |
HIP, HIP/PAP, INGAP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3 |
Location: |
Chr.2:79157006-79159754 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REG3A | NM_002580.2 | NP_002571.1 | ||
| NM_138937.2 | NP_620354.1 | |||
| NM_138938.2 | NP_620355.1 | |||
| AK291973.1 | ||||
| AY544128.1 | AAT11159.1 | |||
| AY544130.1 | AAT11161.1 | |||
| BC036776.1 | AAH36776.1 | |||
| BQ777112.1 | ||||
| D13510.1 | BAA02728.1 | |||
| M84337.1 | AAA36415.1 | |||
| S51768.1 | AAB24642.1 | |||
| X68641.1 | CAA48605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584761 | Chr.2:78317894 - 79203502 on Build GRCh38 | Loss |
|
| dgv1135e212 | Chr.2:79147050 - 79171241 on Build GRCh38 | Loss |
|
| esv3892770 | Chr.2:78806791 - 79322248 on Build GRCh38 | Gain |
|
| nsv522260 | Chr.2:78312540 - 79523465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map