Assay Details
Target Gene Details
Entrez Gene ID: | 11034 |
Gene Name: | destrin, actin depolymerizing factor |
Gene Aliases: |
ACTDP, ADF, HEL32, bA462D18.2 |
Location: |
Chr.20:17569173-17608242 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSTN | NM_001011546.1 | NP_001011546.1 | ||
| NM_006870.3 | NP_006861.1 | |||
| XM_011529142.1 | XP_011527444.1 | |||
| XM_011529143.1 | XP_011527445.1 | |||
| XM_011529144.1 | XP_011527446.1 | |||
| AK302338.1 | ||||
| AK308562.1 | ||||
| AK312645.1 | ||||
| AK315994.1 | ||||
| BC009477.2 | AAH09477.1 | |||
| BX647319.1 | ||||
| CR457042.1 | CAG33323.1 | |||
| CR541956.1 | CAG46754.1 | |||
| EU794606.1 | ||||
| S65738.1 | AAB28361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3311 | Chr.20:17594661 - 17627255 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map