Assay Details
Target Gene Details
Entrez Gene ID: | 10117 |
Gene Name: | enamelin |
Gene Aliases: |
ADAI, AI1C, AIH2 |
Location: |
Chr.4:70627471-70646819 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENAM | NM_031889.2 | 9 | 3032 | NP_114095.2 |
| XM_006714056.3 | 8 | 4781 | XP_006714119.1 | |
| AF125373.1 | 9 | 3032 | AAG43242.1 | |
| BC117308.1 | 8 | 2821 | ||
| BC117310.1 | 8 | 2821 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map